Assay Details
| Gene Symbol: PCDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745925.4 | XP_002745971.2 | 2 - 3 | 3362 | 76 | |
| XM_017970634.1 | XP_017826123.1 | 3 - 4 | 3483 | 76 | ||
More Information
Set Membership: |
|

PCDH7
Genomic Map