Assay Details
| Gene Symbol: HSPA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744205.4 | XP_002744251.1 | 10 - 11 | 1902 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSPA9| Gene Symbol: HSPA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744205.4 | XP_002744251.1 | 10 - 11 | 1902 | 98 | |
Set Membership: |
|