Assay Details
| Gene Symbol: PPP2R5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760545.4 | XP_002760591.1 | 2 - 3 | 1127 | 120 | |
| XM_008985253.2 | XP_008983501.1 | 2 - 3 | 1127 | 120 | ||
More Information
Set Membership: |
|

PPP2R5A
Genomic Map