Assay Details
| Gene Symbol: STK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970982.1 | XP_017826471.1 | 5 - 6 | 1111 | 56 | |
| XM_017970983.1 | XP_017826472.1 | 4 - 5 | 1200 | 56 | ||
More Information
Set Membership: |
|

STK19
Genomic Map