Assay Details
| Gene Symbol: NTM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007153.2 | XP_009005401.2 | 4 - 5 | 1111 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NTM| Gene Symbol: NTM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007153.2 | XP_009005401.2 | 4 - 5 | 1111 | 67 | |
Set Membership: |
|