Assay Details
| Gene Symbol: DPYSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979086.2 | XP_008977334.1 | 11 - 12 | 2069 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPYSL2| Gene Symbol: DPYSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979086.2 | XP_008977334.1 | 11 - 12 | 2069 | 85 | |
Set Membership: |
|