Assay Details
| Gene Symbol: PKNOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007057.1 | XP_009005305.1 | 9 - 10 | 1125 | 57 | |
| XM_009007059.2 | XP_009005307.1 | 10 - 11 | 1170 | 57 | ||
| XM_009007060.2 | XP_009005308.1 | 8 - 9 | 986 | 57 | ||
More Information
Set Membership: |
|

PKNOX2
Genomic Map