Assay Details
| Gene Symbol: MARCH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976560.1 | XP_017832049.1 | 3 - 4 | 1234 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARCH9| Gene Symbol: MARCH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976560.1 | XP_017832049.1 | 3 - 4 | 1234 | 72 | |
Set Membership: |
|