Assay Details
| Gene Symbol: CASP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000457.2 | XP_008998705.2 | 6 - 7 | 844 | 70 | |
| XM_017974346.1 | XP_017829835.1 | 6 - 7 | 844 | 70 | ||
More Information
Set Membership: |
|

CASP9
Genomic Map