Assay Details
| Gene Symbol: SLC33A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759466.2 | XP_002759512.1 | 2 - 3 | 1559 | 82 | |
| XM_008983620.2 | XP_008981868.1 | 2 - 3 | 1559 | 82 | ||
| XM_017965736.1 | XP_017821225.1 | 2 - 3 | 1559 | 82 | ||
More Information
Set Membership: |
|

SLC33A1
Genomic Map