Assay Details
| Gene Symbol: ENTPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017979186.1 | XP_017834675.1 | 7 - 8 | 1184 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENTPD1| Gene Symbol: ENTPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017979186.1 | XP_017834675.1 | 7 - 8 | 1184 | 68 | |
Set Membership: |
|