Assay Details
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755541.3 | XP_002755587.1 | 2 - 3 | 791 | 67 | |
| XM_009008487.2 | XP_009006735.1 | 1 - 2 | 6428 | 67 | ||
More Information
Set Membership: |
|

OVOL1
Genomic Map