Assay Details
| Gene Symbol: ALOX5AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806884.4 | XP_002806930.1 | 2 - 3 | 253 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALOX5AP| Gene Symbol: ALOX5AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806884.4 | XP_002806930.1 | 2 - 3 | 253 | 70 | |
Set Membership: |
|