Assay Details
| Gene Symbol: MRPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759867.3 | XP_002759913.1 | 2 - 3 | 279 | 63 | |
| XM_003735111.2 | XP_003735159.1 | 2 - 3 | 340 | 63 | ||
More Information
Set Membership: |
|

MRPS21
Genomic Map