Assay Details
| Gene Symbol: PRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988946.2 | XP_008987194.1 | 6 - 7 | 988 | 67 | |
| XM_008988947.2 | XP_008987195.1 | 6 - 7 | 971 | 67 | ||
More Information
Set Membership: |
|

PRPS2
Genomic Map