Assay Details
| Gene Symbol: RXRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760442.4 | XP_002760488.1 | 12 - 13 | 2667 | 60 | |
| XM_008985111.2 | XP_008983359.1 | 8 - 9 | 1458 | 60 | ||
| XM_017966115.1 | XP_017821604.1 | 15 - 16 | 2683 | 60 | ||
| XM_017966116.1 | XP_017821605.1 | 13 - 14 | 2251 | 60 | ||
| XM_017966117.1 | XP_017821606.1 | 15 - 16 | 2501 | 60 | ||
| XM_017966119.1 | XP_017821608.1 | 13 - 14 | 1897 | 60 | ||
| GenBank mRNA | EF215260.1 | - | 3 - 4 | 234 | 60 | |
More Information
Set Membership: |
|

RXRG
Genomic Map