Assay Details
| Gene Symbol: PRKAR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748661.4 | XP_002748707.1 | 8 - 9 | 942 | 81 | |
| XM_008997645.2 | XP_008995893.1 | 8 - 9 | 1126 | 81 | ||
| XM_017972761.1 | XP_017828250.1 | 8 - 9 | 968 | 81 | ||
More Information
Set Membership: |
|

PRKAR1A
Genomic Map