Assay Details
| Gene Symbol: AMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755043.3 | XP_002755089.1 | 13 - 14 | 2547 | 63 | |
| XM_009007654.2 | XP_009005902.1 | 13 - 14 | 3260 | 63 | ||
| XM_009007655.2 | XP_009005903.1 | 13 - 14 | 2521 | 63 | ||
| XM_009007656.2 | XP_009005904.1 | 13 - 14 | 2744 | 63 | ||
| XM_009007658.2 | XP_009005906.1 | 12 - 13 | 5327 | 63 | ||
| XM_009007659.1 | XP_009005907.1 | 11 - 12 | 1817 | 63 | ||
More Information
Set Membership: |
|

AMPD3
Genomic Map