Assay Details
| Gene Symbol: CPT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980788.2 | XP_008979036.1 | 11 - 12 | 2158 | 84 | |
| XM_008980803.2 | XP_008979051.1 | 11 - 12 | 1412 | 84 | ||
| XM_008980805.2 | XP_008979053.1 | 11 - 12 | 1489 | 84 | ||
| XM_008980815.2 | XP_008979063.1 | 11 - 12 | 2271 | 84 | ||
More Information
Set Membership: |
|

CPT1B
Genomic Map