Assay Details
| Gene Symbol: LOC100387916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758966.3 | XP_002759012.1 | 15 - 16 | 2264 | 82 | |
| XM_008982729.2 | XP_008980977.1 | 16 - 17 | 1800 | 82 | ||
| XM_008982730.2 | XP_008980978.1 | 15 - 16 | 1455 | 82 | ||
| XM_008982735.2 | XP_008980983.1 | 14 - 15 | 1692 | 82 | ||
| XM_008982736.2 | XP_008980984.1 | 13 - 14 | 1347 | 82 | ||
| XM_017965376.1 | XP_017820865.1 | 16 - 17 | 1514 | 82 | ||
| XM_017965377.1 | XP_017820866.1 | 15 - 16 | 1443 | 82 | ||
| XM_017965378.1 | XP_017820867.1 | 15 - 16 | 1505 | 82 | ||
| XM_017965379.1 | XP_017820868.1 | 12 - 13 | 1062 | 82 | ||
More Information
Set Membership: |
|

LOC100387916
Genomic Map