Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003624.1 | XP_009001872.1 | 5 - 6 | 483 | 133 | |
| XM_009003625.1 | XP_009001873.1 | 3 - 4 | 408 | 133 | ||
| XM_009003626.1 | XP_009001874.1 | 4 - 5 | 381 | 133 | ||
| XM_009003627.2 | XP_009001875.1 | 4 - 5 | 982 | 133 | ||
| XM_009003630.2 | XP_009001878.1 | 6 - 7 | 951 | 133 | ||
| XM_009003631.2 | XP_009001879.1 | 5 - 6 | 910 | 133 | ||
| XM_009003632.2 | XP_009001880.1 | 5 - 6 | 451 | 133 | ||
| XM_017975988.1 | XP_017831477.1 | 3 - 4 | 332 | 133 | ||
| XM_017975989.1 | XP_017831478.1 | 5 - 6 | 2224 | 133 | ||
| XM_017975990.1 | XP_017831479.1 | 5 - 6 | 543 | 133 | ||
More Information
Set Membership: |
|

YAF2
Genomic Map