Assay Details
| Gene Symbol: METAP1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806908.3 | XP_002806954.2 | 2 - 3 | 2101 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

METAP1D| Gene Symbol: METAP1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806908.3 | XP_002806954.2 | 2 - 3 | 2101 | 65 | |
Set Membership: |
|