Assay Details
| Gene Symbol: DCAF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743102.3 | XP_002743148.1 | 6 - 7 | 1410 | 68 | |
| XM_008999675.2 | XP_008997923.1 | 5 - 6 | 1212 | 68 | ||
More Information
Set Membership: |
|

DCAF10
Genomic Map