Assay Details
| Gene Symbol: PLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756960.4 | XP_002757006.2 | 2 - 3 | 336 | 73 | |
| XM_008979334.2 | XP_008977582.2 | 2 - 3 | 339 | 73 | ||
More Information
Set Membership: |
|

PLPP5
Genomic Map