Assay Details
| Gene Symbol: PARVA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755060.4 | XP_002755106.3 | 6 - 7 | 1100 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PARVA| Gene Symbol: PARVA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755060.4 | XP_002755106.3 | 6 - 7 | 1100 | 63 | |
Set Membership: |
|