Assay Details
| Gene Symbol: NOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003471.1 | XP_009001719.1 | 8 - 9 | 1378 | 68 | |
| XM_009003473.1 | XP_009001721.1 | 8 - 9 | 1366 | 68 | ||
| XM_017975880.1 | XP_017831369.1 | 7 - 8 | 881 | 68 | ||
| XM_017975881.1 | XP_017831370.1 | 7 - 8 | 869 | 68 | ||
More Information
Set Membership: |
|

NOP2
Genomic Map