Assay Details
| Gene Symbol: SPPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004790.2 | XP_009003038.1 | 9 - 10 | 1926 | 64 | |
| XM_017976429.1 | XP_017831918.1 | 10 - 11 | 2033 | 64 | ||
| XM_017976430.1 | XP_017831919.1 | 9 - 10 | 1114 | 64 | ||
More Information
Set Membership: |
|

SPPL3
Genomic Map