Assay Details
| Gene Symbol: HNF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997016.2 | XP_008995264.1 | 6 - 7 | 1555 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HNF1B| Gene Symbol: HNF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997016.2 | XP_008995264.1 | 6 - 7 | 1555 | 59 | |
Set Membership: |
|