Assay Details
| Gene Symbol: RAB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743263.4 | XP_002743309.1 | 5 - 6 | 794 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB14| Gene Symbol: RAB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743263.4 | XP_002743309.1 | 5 - 6 | 794 | 83 | |
Set Membership: |
|