Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756053.4 | XP_002756099.1 | 4 - 5 | 586 | 113 | |
| XM_009009426.2 | XP_009007674.1 | 4 - 5 | 548 | 113 | ||
More Information
Set Membership: |
|

CD2BP2
Genomic Map