Assay Details
| Gene Symbol: TYRO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753517.4 | XP_002753563.1 | 10 - 11 | 1424 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TYRO3| Gene Symbol: TYRO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753517.4 | XP_002753563.1 | 10 - 11 | 1424 | 61 | |
Set Membership: |
|