Assay Details
| Gene Symbol: RGL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967806.1 | XP_017823295.1 | 11 - 12 | 1426 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGL3| Gene Symbol: RGL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967806.1 | XP_017823295.1 | 11 - 12 | 1426 | 78 | |
Set Membership: |
|