Assay Details
| Gene Symbol: SNUPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005286.1 | XP_009003534.1 | 5 - 6 | 635 | 98 | |
| XM_009005287.2 | XP_009003535.1 | 5 - 6 | 587 | 98 | ||
| XM_009005288.2 | XP_009003536.1 | 5 - 6 | 848 | 98 | ||
| XM_017976767.1 | XP_017832256.1 | 5 - 6 | 947 | 98 | ||
More Information
Set Membership: |
|

SNUPN
Genomic Map