Assay Details
Gene Symbol: THBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744826.4 | XP_002744872.1 | 21 - 22 | 3011 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: THBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744826.4 | XP_002744872.1 | 21 - 22 | 3011 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |