Assay Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003734524.3 | XP_003734572.1 | 8 - 9 | 1216 | 73 | |
XM_009010338.2 | XP_009008586.1 | 8 - 9 | 1333 | 73 | ||
XM_009010339.2 | XP_009008587.1 | 8 - 9 | 1333 | 73 | ||
XM_017963195.1 | XP_017818684.1 | 9 - 10 | 871 | 73 | ||
XM_017963196.1 | XP_017818685.1 | 8 - 9 | 719 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
