Assay Details
| Gene Symbol: TBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732732.3 | XP_003732780.1 | 2 - 3 | 435 | 113 | |
| XM_008995095.1 | XP_008993343.1 | 3 - 4 | 511 | 113 | ||
| XM_008995096.1 | XP_008993344.1 | 3 - 4 | 615 | 113 | ||
More Information
Set Membership: |
|

TBPL1
Genomic Map