Assay Details
| Gene Symbol: HM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747368.3 | XP_002747414.1 | 3 - 4 | 524 | 86 | |
| XM_002747369.3 | XP_002747415.1 | 3 - 4 | 524 | 86 | ||
| XM_008995584.2 | XP_008993832.1 | 3 - 4 | 524 | 86 | ||
| XM_017971863.1 | XP_017827352.1 | 4 - 5 | 651 | 86 | ||
| XM_017971864.1 | XP_017827353.1 | 4 - 5 | 651 | 86 | ||
| XM_017971865.1 | XP_017827354.1 | 4 - 5 | 651 | 86 | ||
More Information
Set Membership: |
|

HM13
Genomic Map