Assay Details
| Gene Symbol: GARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751430.4 | XP_002751476.1 | 7 - 8 | 1113 | 69 | |
| XM_017975219.1 | XP_017830708.1 | 7 - 8 | 1113 | 69 | ||
More Information
Set Membership: |
|

GARS
Genomic Map