Assay Details
| Gene Symbol: CAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993922.2 | XP_008992170.1 | 12 - 13 | 2585 | 93 | |
| XM_017970887.1 | XP_017826376.1 | 12 - 13 | 1370 | 93 | ||
| XM_017970888.1 | XP_017826377.1 | 12 - 13 | 1370 | 93 | ||
More Information
Set Membership: |
|

CAP2
Genomic Map