Assay Details
| Gene Symbol: LTC4S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744461.3 | XP_002744507.2 | 2 - 3 | 859 | 61 | |
| XM_008989486.2 | XP_008987734.1 | 2 - 3 | 859 | 61 | ||
| XM_008989503.2 | XP_008987751.1 | 2 - 3 | 859 | 61 | ||
More Information
Set Membership: |
|

LTC4S
Genomic Map