Assay Details
| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757938.4 | XP_002757984.2 | 2 - 3 | 503 | 68 | |
| XM_008980978.2 | XP_008979226.1 | 2 - 3 | 503 | 68 | ||
More Information
Set Membership: |
|

PPM1G
Genomic Map