Assay Details
| Gene Symbol: PSMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972464.1 | XP_017827953.1 | 2 - 3 | 199 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMC5| Gene Symbol: PSMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972464.1 | XP_017827953.1 | 2 - 3 | 199 | 70 | |
Set Membership: |
|