Assay Details
| Gene Symbol: TFAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746236.3 | XP_002746282.1 | 2 - 3 | 1257 | 58 | |
| XM_008993986.2 | XP_008992234.1 | 2 - 3 | 605 | 58 | ||
More Information
Set Membership: |
|

TFAP2A
Genomic Map