Assay Details
| Gene Symbol: PLCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759693.4 | XP_002759739.2 | 1 - 2 | 405 | 73 | |
| XM_017965891.1 | XP_017821380.1 | 1 - 2 | 373 | 73 | ||
More Information
Set Membership: |
|

PLCL2
Genomic Map