Assay Details
| Gene Symbol: CYLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760943.3 | XP_002760989.1 | 3 - 4 | 1159 | 73 | |
| XM_008985856.1 | XP_008984104.1 | 4 - 5 | 1044 | 73 | ||
| XM_017966924.1 | XP_017822413.1 | 4 - 5 | 1117 | 73 | ||
| XM_017966925.1 | XP_017822414.1 | 4 - 5 | 1102 | 73 | ||
| XM_017966926.1 | XP_017822415.1 | 4 - 5 | 1098 | 73 | ||
| XM_017966927.1 | XP_017822416.1 | 3 - 4 | 1140 | 73 | ||
| XM_017966928.1 | XP_017822417.1 | 4 - 5 | 1044 | 73 | ||
| XM_017966929.1 | XP_017822418.1 | 3 - 4 | 1156 | 73 | ||
More Information
Set Membership: |
|

CYLD
Genomic Map