Assay Details
| Gene Symbol: AMFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760964.3 | XP_002761010.1 | 4 - 5 | 841 | 95 | |
| XM_017966783.1 | XP_017822272.1 | 4 - 5 | 841 | 95 | ||
More Information
Set Membership: |
|

AMFR
Genomic Map