Assay Details
| Gene Symbol: SRSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757833.4 | XP_002757879.1 | 3 - 4 | 622 | 62 | |
| XM_003734752.3 | XP_003734800.1 | 3 - 4 | 623 | 62 | ||
| XM_008980832.2 | XP_008979080.1 | 3 - 4 | 623 | 62 | ||
| XM_008980833.2 | XP_008979081.1 | 3 - 4 | 623 | 62 | ||
More Information
Set Membership: |
|

SRSF7
Genomic Map