Assay Details
| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735082.3 | XP_003735130.1 | 4 - 5 | 626 | 106 | |
| XM_017965956.1 | XP_017821445.1 | 4 - 5 | 465 | 106 | ||
| XM_017965957.1 | XP_017821446.1 | 4 - 5 | 366 | 106 | ||
More Information
Set Membership: |
|

EXOG
Genomic Map