Assay Details
| Gene Symbol: NPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753317.4 | XP_002753363.1 | 3 - 4 | 806 | 78 | |
| XM_009005218.2 | XP_009003466.1 | 3 - 4 | 806 | 78 | ||
| XM_009005219.1 | XP_009003467.1 | 2 - 3 | 460 | 78 | ||
| XM_009005220.1 | XP_009003468.1 | 2 - 3 | 460 | 78 | ||
More Information
Set Membership: |
|

NPTN
Genomic Map