Assay Details
| Gene Symbol: ACTR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991932.2 | XP_008990180.1 | 3 - 4 | 412 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACTR1B| Gene Symbol: ACTR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991932.2 | XP_008990180.1 | 3 - 4 | 412 | 77 | |
Set Membership: |
|